ContactUsLinkComponent
Test Kits, model FF-2 for freshwater aquaculture
Test Kits, model FF-2 for freshwater aquaculture
Catalog # HACH243001
Supplier:  Hach
CAS Number:  
undefined
Test Kits, model FF-2 for freshwater aquaculture
Catalog # HACH243001
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Model FF-2, Ten-parameter test kit for freshwater fish production
Specifications
About this item
Maintaining good water quality is essential and this kit utilizes drop count titration and color disc methods to deliver the fast, accurate results. Allows measurement of ten critical freshwater parameters using accurate Digital Titrator titration and continuous colour disc colorimetry.
Delivery: Includes 100 ml bottles of deionised water, twenty three reagents, one flask, clippers, one graduated cylinder, one color comparator box, one digital titrator, one delivery tube, two plastic tubes, one thermometer, one BOD bottle, three colour discs, a manual, and a carrying case.