Orders LinkContactUsLinkComponent
Storage Boxes
Storage Boxes
Supplier:   SEMADENI
Storage Boxes
Supplier:  SEMADENI
Certificates
3 Options Available Below
Product Details & Documents
PP, translucent.
  • Very stable
  • Good chemical resistance
  • Good temperature resistance
Multipurpose boxes for a wide range of uses.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • SEMA5971
    12 compartments
    12 compartments
    250×170×46 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.50
    SFr. 19.50
    1 * 1 ST
       SFr. 19.50
  • SEMA5972
    13 compartments
    13 compartments
    250×170×46 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.50
    SFr. 19.50
    1 * 1 ST
       SFr. 19.50
  • SEMA5973
    24 compartments
    24 compartments
    335×225×55 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 26.90
    SFr. 26.90
    1 * 1 ST
       SFr. 26.90
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..