SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Digestion Systems
Kjeldahl Tablets and Reagents
Kjeldahl tablets, Kjeltab
Kjeldahl tablets, Kjeltab
Catalog # GERH12-0326
Supplier:
GERHARDT
CAS Number:
Expand image
Kjeldahl tablets, Kjeltab
Catalog # GERH12-0326
Supplier:
GERHARDT
CAS Number:
Specifications
Pk:
1.000
Description:
Kjeltab ST, 3,5 g
Ø:
16 mm
Contents:
K
2
SO
4
3,5 g; Se 0,0035 g
Specifications
About this item
Product Details
Product Details