Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Stirrers
Stirring Rods
Stirring rods
Stirring rods
Stirring rods
Catalog # HACH177001
Supplier:
Hach
Expand image
Stirring rods
Catalog # HACH177001
Supplier:
Hach
Specifications
Ø:
5 mm
Description:
Stirring rod
Length:
150 mm
Material:
Glass
Pk:
3
Specifications
About this item
Product Details
Documentation
Product Details
Documentation