Order Entry
Switzerland
ContactUsLinkComponent
 
Centrifugal filtration units, Vivaspin® Turbo 4
 :  Sartorius
 
undefined
Centrifugal filtration units, Vivaspin® Turbo 4
 :  Sartorius

 

  • Catalog No:
  • 518-0096
  • 518-0095
  • 518-0098
  • 518-0097
  • 518-0103
  • 518-0102
  • 518-0094
  • 518-0099
  • 518-0101
  • 518-0100
  • 518-0092
  • 518-0093
  • Filtration area:
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • 3,2 cm²
  • Max. RCF - swing-out rotor:
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 100 kD: 3000 ×g
  • For 100 kD: 3000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • For 3 kD - 50 kD: 4000 ×g
  • Max. RCF - fixed angle rotor:
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 100 kD: 5000 ×g (25°)
  • For 100 kD: 5000 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • For 3 kD - 50 kD: 7500 ×g (25°)
  • Dead volume:
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • <10 µl
  • Ø×H:
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm
  • 14,6×75,5 mm

 

Frequently Bought Together