ContactUsLinkComponent
Phosphorus (Reactive) TNT reagent set
Phosphorus (Reactive) TNT reagent set
Catalog # 705-2049
Supplier:  Hach
CAS Number:  
undefined
Phosphorus (Reactive) TNT reagent set
Catalog # 705-2049
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    Molybdovanadate
  • Pk:
    1 SET
  • Description:
    Phosphorus TNT reagent set, High range
  • For unit:
    DR900
  • Tests per kit:
    50
  • Range:
    1,0 - 100,0 mg/L PO₄
Specifications
About this item
Set used for determination of reactive phosphorus (ortho-phosphate) by the molybdovanadate Test 'N Tube™ method.
  • Test 'N Tube products provide safe, convenient testing
  • Easy ordering
Delivery: Set includes vials and reagents for up to 50 tests (dependent upon the number of reagent blank tests performed).