ContactUsLinkComponent
Nitrite test kit, colour disc, NI-6
Nitrite test kit, colour disc, NI-6
Catalog # HACH224000
Supplier:  Hach
CAS Number:  
undefined
Nitrite test kit, colour disc, NI-6
Catalog # HACH224000
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Nitrite test kit, HR, NI-6
  • Range:
    0 - 80 mg/l NO₂
    0 - 1600 mg/l NO₂
Specifications
About this item
The kit uses the colour disc or ferrous sulphate method to measure nitrogen as nitrite NO²⁻.
  • Simple, cost effective
  • Portable test kit for analysis in the field
Delivery: Kit includes colour disc, comparator box and tubes, reagents, deionised dilution water, instructions and carrying case.