Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Nessler reagent

Supplier:   Hach
Nessler reagent
Supplier:  Hach
Certificates
Nessler reagent

2 Options Available Below

Product Details & Documents

The reagent is used for ammonia nitrogen determination in waste water by the Nessler method

Synonyms:  Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution
  • CAS Number:7783-33-7
  • MDL Number:MFCD00049656
Specifications for Product Family

Specification Test Results

  • Range:
    0.02-2.50 mg/L NH3-N
Product Family Options

Product Information