Order Entry
Switzerland
ContactUsLinkComponent
Iron(III) chloride solution in sulphuric acid
Corrosive
Iron(III) chloride solution in sulphuric acid
Catalog # HACH204253
Supplier:  Hach
CAS Number:  
undefined
Corrosive
Iron(III) chloride solution in sulphuric acid
Catalog # HACH204253
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :162.21 g/mol
UN:3264
ADR:8,III
 :GHS05
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005