Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Conductivity Standards
Conductivity calibration solutions, YSI
Conductivity calibration solutions, YSI
Conductivity calibration solutions, YSI
Catalog # YSIA060660
Supplier:
YSI ENVIRONMENTAL
CAS Number:
Expand image
Conductivity calibration solutions, YSI
Catalog # YSIA060660
Supplier:
YSI ENVIRONMENTAL
CAS Number:
Specifications
Pk:
8
Description:
Conductivity standard
Conductivity:
50 000 µS/cm
Specifications
About this item
Product Details
Product Details