Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, WHEATON®
Dropping bottles, LDPE, WHEATON®
Supplier:   DWK Life Sciences
Dropping bottles, LDPE, WHEATON®
Supplier:  DWK Life Sciences
Certificates
Dropping bottles, LDPE, WHEATON®
Dropping bottles, LDPE, WHEATON®
4 Options Available Below
Product Details & Documents
LDPE, translucent, with tip and cap.
  • Tip dispenses a stream of liquid or non-controlled drop
  • Flexible for easy handling
  • Good chemical and temperature resistance
Packaging: Dropper bottles, tips and caps are packaged separately.
Product Family Options
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 7 ml
    63 mm
    19 mm
    15-415
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 421.00 SFr.
    421.00 SFr.
    1 * 144 ST
       421.00 SFr.
  • 15 ml
    78 mm
    25 mm
    15-415
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 453.00 SFr.
    453.00 SFr.
    1 * 144 ST
       453.00 SFr.
  • 30 ml
    85 mm
    32 mm
    20-410
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 500.00 SFr.
    500.00 SFr.
    1 * 144 ST
       500.00 SFr.
  • 60 ml
    101 mm
    39 mm
    20-410
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 535.00 SFr.
    535.00 SFr.
    1 * 144 ST
       535.00 SFr.