ContactUsLinkComponent
Test kits, tannin and lignin
Test kits, tannin and lignin
Catalog # HACH193701
Supplier:  Hach
CAS Number:  
undefined
Test kits, tannin and lignin
Catalog # HACH193701
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Tannin/lignin test kit, model TA-3
  • Range:
    0 - 15 mg/l
    0 - 150 mg/l
Specifications
About this item
Designed to analyse tannin and lignin together and report result as mg/l tannic acid.
  • Simple, economical colour disc method
  • Easy-to-carry test kit with quick results
Delivery: Kit includes colour disc, comparator box and tubes, reagents, deionized water, mixing bottle, funnel, filter papers, instructions and carrying case.