ContactUsLinkComponent
Visual colour comparator kits, manganese
Visual colour comparator kits, manganese
Catalog # HACH2350800
Supplier:  Hach
CAS Number:  
undefined
Visual colour comparator kits, manganese
Catalog # HACH2350800
Supplier:  Hach
CAS Number:  
Specifications
  • Chemical method:
    PAN
  • Pk:
    1 KIT
  • Description:
    Manganese colour disc test kit, model MN-PAN
  • Range:
    0 - 0,70 mg/l
Specifications
About this item
A simple, economical and rapid visual colour determination kit.
Delivery: Kit includes colour disc, comparator box and tubes, reagents, demineraliser bottle, mixing bottles, instructions and carrying case.