ContactUsLinkComponent
Molybdate test kit, MO-LR
  HACH2359300
 :  Hach
 :  
undefined
Molybdate test kit, MO-LR
  HACH2359300
 :  Hach
 :  
  • Pk:
    1 KIT
  • Description:
    Molybdate test kit, MO-LR
  • Range:
    0 - 3,0 mg/l Mo
Molybdates are used to inhibit corrosion in industrial and utility water systems. The reagents in the kit reacts with molybdate to form a blue colour, which is measured with a colour disc.
  • Easy to use colour disc
  • Portable test kit for on-site measurement
  • Effectively monitors the levels of molybdate corrosion inhibitors
 : The kit includes two reagents, one color comparator box, one color disc, two stoppers, instructions, and a carrying case.