ContactUsLinkComponent
Visual comparator test kit, ethylene glycol
Visual comparator test kit, ethylene glycol
  HACH2186400
 :  Hach
 :  
undefined
Visual comparator test kit, ethylene glycol
  HACH2186400
 :  Hach
 :  
  • Pk:
    1 KIT
  • Description:
    Ethylene glycol, EG-1 kit
  • Range:
    0 - 150 mg/l
Ethylene glycol (anti-freeze) contamination of automotive lubricants is an indication of a cracked engine block. Glycol presence also can indicate leaks in cooling systems. This test kit model EG-1 presents a purple colour if ethylene glycol and other 1,2 glycols are present in oil or water.
 : Kit includes sample pipettes, vials and reagents for 25 tests, standard solution, instructions and carrying case.