Lysates, human
About this item
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
- Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.
9 Options Available Below
Product Details & Documents
Whole cell lysates are derived from cell lines tissues using highly refined extraction protocols to ensure quality, protein integrity and high reproducibility. Lysates are prepared in denaturing buffer without dissociating agents.
- Liquid
Whole cell lysates are ready to use for SDS-PAGE and Western blotting purposes. All extracts are tested by SDS-PAGE using 4 to 20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Each vial contains 1mg/ml of protein in SDS-PAGE sample buffer. The sample buffer consists of 0,05 mM Tris Hcl, 2% SDS, 10% glycerol and 0,1% bromophenol blue at pH 6,8.
Documents
Product Family Options
Product Information
- PkAvailabilityPrice
- ROCKW09-000-360MCF-7 whole cell lysate
- ROCKW09-000-366MCF-7 whole cell lysate, hydrogen peroxide stimulatedMCF-7 whole cell lysate, hydrogen peroxide stimulated500 µGLog in for availability1 * 500 µGSFr. 376.00
- ROCKW09-000-361A431 whole cell lysate
- ROCKW09-000-362A431 whole cell lysate - EGF stimulated
- ROCKW09-000-363Hep2 whole cell lysate
- ROCKW09-000-364HeLa whole cell lysate
- ROCKW09-000-365HEK293, whole cell lysate
- ROCKW09-001-370Jurkat whole cell lysate
- ROCKW09-001-GY4U251 Whole Cell Lysate