Reagent reservoirs
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
Disposable reagent reservooirs made of resistant and durable PET.
- Pour-off spouts on all four corners
- Wide base and reinforced sidewalls avoid spills and provide stability
- Sterilised by gamma irradiation
- 25 ml reservoir with divider accommodates eight channel pipettes on one side and one to four channel pipettes on the other; divider is also convenient for adding controls
- Optional coolers are available which keep the reagents at ~5 °C for approximately 3 hours
Frequently Bought Together