Order Entry
Switzerland
ContactUsLinkComponent
Ammonium dihydrogen phosphate 100 g/l in water phosphate modifier for graphite furnace AAS, Supelco®
Ammonium dihydrogen phosphate 100 g/l in water phosphate modifier for graphite furnace AAS, Supelco®
Catalog # 1.07290.0050
Supplier:  Merck
CAS Number:  
Ammonium dihydrogen phosphate 100 g/l in water phosphate modifier for graphite furnace AAS, Supelco®
Catalog # 1.07290.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Concentration ((NH₄)H₂PO₄):
    100 ± 2 g/l

Specification Test Results

About this item

 :
Boiling Pt:100 °C
Density:1.052 g/cm3 (20 °C)
 :M-Clarity™ quality level = MQ100
 :115.03 g/mol
Storage Temperature:Ambient
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396