Precision Clean® II ultra clear cleanroom tubing
Nome del fornitore: FISHER CONTAINER
Certificati
Informazioni su questo articolo
This clear Polyethylene tubing features an inner surface cleanliness level suitable for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom conditions.
Ordering information: Individually wrapped and shipped with special plastic cores to prevent particulate contamination.
Frequently Bought Together