Order Entry
Switzerland
ContactUsLinkComponent
 
D-(+)-Glucose monohydrate, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 1910-05
Supplier:  Avantor
CAS Number:  
undefined
D-(+)-Glucose monohydrate, powder, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 1910-05
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 0.005 %
  • Insoluble Matter:
    ≤ 0.005 %
  • Residue after Ignition:
    ≤ 0.015 %
  • Chloride (Cl):
    ≤ 0.005 %
  • Sulfate and Sulfite (as SO₄):
    ≤ 0.005 %
  • Starch:
    Passes Test
  • Arsenic (As):
    ≤ 0.200 ppm
  • Trace Impurities - ACS - Heavy Metals (as Pb):
    ≤ 5 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 5 ppm

Specification Test Results

About this item

 :
 :198.17 g/mol
Melting Pt:83 °C
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:14431-43-7
 :5996-10-1
Formula:C₆H₁₂O₆·H₂O
MDL Number:MFCD00149450