Orders LinkContactUsLinkComponent
Insulation stripper
Insulation stripper
Insulation stripper, chrome plated, handles insulated with multi-component grips
Catalog # 238-5410
Supplier:   KNIPEX WERKE
Insulation stripper
Catalog # 238-5410
Supplier:  KNIPEX WERKE
Insulation stripper
Insulation stripper
Insulation stripper, chrome plated, handles insulated with multi-component grips
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Chrome plated, forged, oil-hardened insulation stripper.
  • For single, multiple and fine stranded conductors with plastic or rubber insulation max. Ø 5,0 mm or respectively 10 mm² cable cross section
  • Easy adjustment to the required diameter of solid or stranded wire with knurled screw and lock nut
  • Handles insulated with multi-component grips
Certifications: DIN EN 60900, IEC 60900. VDE-tested
Specifications
  • Description:
    Insulation stripper, chrome plated, handles insulated with multi-component grips