Order Entry
Switzerland
ContactUsLinkComponent
VWR®, Cleanroom Sleeve Protectors
VWR®, Cleanroom Sleeve Protectors
Catalog # 113-7887
Supplier:  VWR Collection
undefined
VWR®, Cleanroom Sleeve Protectors
Catalog # 113-7887
Supplier:  VWR Collection

Specifications

  • Sterile:
    +
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Length:
    430 mm
  • Pk:
    150 PAIR

Specifications

About this item

Tapered sleeve protectors made from breathable SMS. Ideal for use in cleanroom and ESD environments.

  • Elastic at both ends
  • Folded seams
  • Vacuum packed

Weight: 60 g.

Level I acc. IEST-RP-CC03.3. Suitable for use in ISO Class 5 + 6 cleanroom environments.

Certifications: Type 5/6; Cat. III.
Packaging: Individually double-bagged with lining.