Order Entry
Switzerland
ContactUsLinkComponent
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
Catalog # 40011.
Supplier:  Biotium
CAS Number:  
4',6'-Diamidino-2-phenylindole dihydrochloride (DAPI dihydrochloride) used in nuclear staining
Catalog # 40011.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :350.25 g/mol
Storage Temperature:Refrigerator
CAS Number:28718-90-3
Formula:C₁₆H₁₅N₅·2HCl
MDL Number:MFCD00012681