Order Entry
Switzerland
ContactUsLinkComponent
 
(±)-1,2-Propanediol ≥99.5% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # U510-07
Supplier:  Avantor
CAS Number:  
undefined
(±)-1,2-Propanediol ≥99.5% (dry basis), BAKER ANALYZED® ACS, J.T.Baker®
Catalog # U510-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle in a single shipper
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    = 99.5 %
  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • ACS - Assay (CH₃CHOHCH₂OH) (corrected for water):
    ≥99.5 %
  • ACS - Color (APHA):
    ≤10
  • ACS - Residue after Ignition:
    ≤0.005 %
  • ACS - Titrable Acid (meq/g):
    ≤0.0005
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.2 %
  • Chloride (Cl):
    ≤1 ppm

Specification Test Results

About this item

 :
 :76.1 g/mol
Merck Index:13,07947
Boiling Pt:187.6 °C (1013 hPa)
Melting Pt:–59 °C
Density:1.04 g/cm³ (20 °C)
Flash Pt:99 °C
Storage Temperature:Ambient
EINECS:200-338-0
CAS Number:57-55-6
Formula:CH₃CH(OH)CH₂OH
MDL Number:MFCD00064272