Triethanolamine (trolamine) ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Catalog # 9468-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (by GC):≥ 99.0 %
- Residue after Ignition:≤ 0.005 %
- Water (by KF, volumetric):≤ 0.2 %
- Trace Impurities - Iron (Fe):≤ 0.001 %
Specification Test Results
About this item
:
:149.19 g/mol
Merck Index:13,09739
Boiling Pt:335.4 °C (1013 hPa)
Melting Pt:21.6 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:179 °C (closed cup)
Storage Temperature:Ambient
EINECS:203-049-8
CAS Number:102-71-6
Formula:C₆H₁₅NO₃
MDL Number:MFCD00002855