Order Entry
Switzerland
ContactUsLinkComponent
di-Sodium succinate hexahydrate, granular, Macron Fine Chemicals™
di-Sodium succinate hexahydrate, granular, Macron Fine Chemicals™
Catalog # 7980-05
CAS Number:  
undefined
di-Sodium succinate hexahydrate, granular, Macron Fine Chemicals™
Catalog # 7980-05
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Chloride (Cl):
    ≤0.02 %
  • Heavy Metals (as Pb):
    ≤0.002 %
  • pH of 0.4M Solution:
    6.0 - 9.0
  • Solubility:
    Passes Test
  • Substances Reducing Permanganate:
    Passes Test
  • Sulfate (SO₄):
    Passes Test

Specification Test Results

About this item

 :
 :270.14 g/mol
Merck Index:13,08754
Melting Pt:300 °C
Density:1.529 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-778-7
CAS Number:6106-21-4
Formula:NaOOCCH₂CH₂COONa·6H₂O
MDL Number:MFCD00149117