
Sodium hydroxide 1 N in aqueous solution, BAKER ANALYZED®, Laboratory Reagent volumetric solution, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:Filtered through a 0.2 µm filter.
:GHS05
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.995 - 1.005
- Protect from air to avoid absorption of carbon dioxide.:≤5 ppm
- Appearance (Clear, colorless liquid):Passes Test
- Normality (eq/L):0.995 - 1.005
- Chloride (Cl):≤5 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.5 ppm
Specification Test Results
Related Information
Frequently Bought Together