Order Entry
Switzerland
ContactUsLinkComponent
EDTA disodium salt dihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
EDTA disodium salt dihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
EDTA disodium salt dihydrate 99.0-101.0% (dried basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Meets J.P. Chemical Specifications:
  • Meets U.S.P Requirements:
  • Meets B.P. Chemical Specifications:
  • Meets E.P. Chemical Specifications:
  • GMP Manufactured Product:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - pH (1 in 20):
    4.0 - 6.0
  • USP - Loss on Drying:
    8.7 - 11.4 %
  • USP - Calcium (Ca):
    Passes Test
  • USP - Nitrilotriacetic Acid:
    ≤0.1 %
  • USP - Assay (C₁₀H₁₄N₂Na₂O₈) (dried basis):
    99.0 - 101.0 %
  • Endotoxin Concentration (EU/g):
    ≤5
  • EP/BP - Assay:
    98.5 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification D:
    Passes Test
  • EP/BP - Impurity A:
    ≤0.1 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH (1 in 20):
    4.0 - 5.5
  • EP/BP - Iron (Fe):
    ≤80 ppm
  • JP - Assay:
    99.0 - 101.0 %
  • JP - Identification A:
    Passes Test
  • JP - Identification B:
    Passes Test
  • JP - Identification C:
    Passes Test
  • JP - pH of 1% Solution at 25°C:
    4.3 - 4.7
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Cyanide (CN):
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Arsenic (As):
    ≤2 ppm
  • JP - Residue after Ignition:
    37.0 - 39.0 %
  • No Class 1,2,3 or other solvents are used or produced in the:
    37.0 - 39.0 %

Specification Test Results

Related Information

Frequently Bought Together