Order Entry
Switzerland
ContactUsLinkComponent
 
Flammable
Toxic
Health
Methanol ≥99.9% (by GC), UltimAR® for liquid chromatography, for organic residue analysis, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # H488-68
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol ≥99.9% (by GC), UltimAR® for liquid chromatography, for organic residue analysis, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # H488-68
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Appearance:
    Passes Test
  • ACS - Assay (CH₃OH) (by GC, corrected for water):
    ≥99.9 %
  • Carbonyl Compounds - Acetone:
    ≤0.001 %
  • Carbonyl Compounds - Acetaldehyde:
    ≤0.001 %
  • Carbonyl Compounds - Formaldehyde:
    ≤0.001 %
  • Color (APHA):
    ≤5
  • Residue after Evaporation:
    ≤0.00010 %
  • Solubility in H₂O:
    Passes Test
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Substances Reducing Permanganate:
    Passes Test
  • Titrable Acid (meq/g):
    ≤0.0003
  • Titrable Base (meq/g):
    ≤0.0002
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.05 %
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm:
    ≤1.00
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤0.80
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.25
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:
    ≤0.12
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:
    ≤0.05
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 - 280 nm:
    ≤0.005
  • Liquid Chromatography Suitability Absorbance:
    Passes Test
  • HPLC Gradient Elution (No Peak Greater than 25% of Full Scale)(0.005 au):
    Passes Test
  • ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):
    ≤5
  • Sum of the Peaks (ng/L):
    ≤10
  • FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):
    ≤5
  • Sum of the Peaks (µg/L):
    ≤10
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.0 ppb

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595