


Isobutanol ≥99.0%, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:78-83-1
:
Formula:(CH₃)₂CHCH₂OH
MDL Number:MFCD00004740
:74.12 g/mol
Merck Index:13,05148
Boiling Pt:107.9 °C (1013 hPa)
Melting Pt:–108 °C
Density:0.802 g/cm³ (20 °C)
Flash Pt:28 °C
Storage Temperature:Ambient
EINECS:201-148-0
UN:1212
ADR:3,III
:GHS02
:GHS05
:GHS07
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:= 99.0 %
- For Laboratory,Research,or Manufacturing Use:= 0.02 %
- Assay ((CH₃)₂CHCH₂OH) (by GC, corrected for water):≥99.0 %
- Carbonyl Compounds - 2-Butanone:≤0.02 %
- Carbonyl Compounds - Butyraldehyde:≤0.01 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- Solubility in H₂O:Passes Test
- Titrable Acid (meq/g):≤0.0005
- Water (H₂O)(by Karl Fischer titrn):≤0.1 %
Specification Test Results
Frequently Bought Together