Orders LinkContactUsLinkComponent
Screw caps
Screw caps
Supplier:   MURRAY S
Screw caps
Supplier:  MURRAY S
Certificates
5 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • For
    Availability
    Price
  • MURRT001/C1
    Black PP screw cap with foil-faced wads
    Black PP screw cap with foil-faced wads
    11 and 13 mm Ø vials
    Log in for availability
    Log in for availability
    1 * 666 ST
    1 * 666 ST - SFr. 111.00
    SFr. 111.00
    1 * 666 ST
       SFr. 111.00
  • MURRT001/C3
    Black PP screw cap with foil-faced wads
    Black PP screw cap with foil-faced wads
    17 mm Ø vials
    Log in for availability
    Log in for availability
    1 * 399 ST
    1 * 399 ST - SFr. 67.70
    SFr. 67.70
    1 * 399 ST
       SFr. 67.70
  • MURRT001/C5
    Black PP screw cap with foil-faced wads
    Black PP screw cap with foil-faced wads
    20 and 21 mm Ø vials
    Log in for availability
    Log in for availability
    1 * 264 ST
    1 * 264 ST - SFr. 90.20
    SFr. 90.20
    1 * 264 ST
       SFr. 90.20
  • MURRT001/C7
    Black PP screw cap with foil-faced wads
    Black PP screw cap with foil-faced wads
    23 mm Ø vials
    Log in for availability
    Log in for availability
    1 * 190 ST
    1 * 190 ST - SFr. 67.30
    SFr. 67.30
    1 * 190 ST
       SFr. 67.30
  • MURRT002/C3
    Black PP screw cap with foil-faced wads
    Black PP screw cap with foil-faced wads
    28 mm Ø vials
    Log in for availability
    Log in for availability
    1 * 140 ST
    1 * 140 ST - SFr. 25.20
    SFr. 25.20
    1 * 140 ST
       SFr. 25.20
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.