Petri dishes, soda lime silca glass, Corning®
Anbieter: Corning
Zertifikate
Über diesen Artikel
Flat, clear petri dishes are manufactured from soda lime silica glass and will withstand repeated sterilization (wet or dry).
Edges are beaded to provide greater mechanical strength. The bead also provides a means to equally space the side walls of the bottom and cover, reducing capillary action of any condensed moisture within. Dish bottoms feature a triangular enamel reference point for serial dilutions. Cover does not provide airtight seal. Chemical and thermal-resistant.
Caution: These dishes are not designed for rapid thermal shock.
Frequently Bought Together