ContactUsLinkComponent
Immobilised Iminobiotin Resin
Immobilised Iminobiotin Resin
Catalog # 786-599
Supplier:  G-Biosciences
CAS Number:  
undefined
Immobilised Iminobiotin Resin
Catalog # 786-599
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    5 mL
  • Description:
    Iminobiotin resin, immobilised
Specifications
About this item
Immobilised Iminobiotin Resin consists of iminobiotin, a cyclic guanido analog of biotin, covalently coupled to 6% crosslinked agarose. The resin allows for the purification of avidin, streptavidin and Neutravidin and their subsequent gentle elution using non denaturing elution buffers.
  • Biotin binding capacity: 2 mg avidin/ml resin
  • No requirement for strong, denaturing elution buffers
  • Elutes at pH 4,0