Dot blot system, Enhancer™
Catalog # 786-163
Supplier: G-Biosciences
Specifications
- Description:Enhancer™ dot blot system
- Pk:1 KIT
Specifications
About this item
Enhancer™ dot blot system allows for the rapid application of up to 384 protein or nucleic acid samples on to a 12×9 cm membrane for subsequent analysis. Samples are applied to a nitrocellulose or nylon membrane by capillary action, resulting in concentration of sample due to minimal diffusion.
- Increase dot blot sensitivity by 10-fold
- Apply up to 384 protein or nucleic acid samples
- No cross-contamination risk
- No expensive electronic equipment required