Orders LinkContactUsLinkComponent
Gram Negative lysis buffer
Supplier:   G-Biosciences
Gram Negative lysis buffer
Supplier:  G-Biosciences
Certificates
Gram Negative lysis buffer
Gram Negative lysis buffer
4 Options Available Below
Product Details & Documents
Gram-negative lysis buffer is an extraction buffer for soluble proteins from Gram-negative bacteria. It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins. This buffer has been tested for use with several widely used bacteria including E.coli strains.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • 786-566
    Gram negative lysis buffer
    Gram negative lysis buffer
    125 mL
    Log in for availability
    Log in for availability
    1 * 125 mL
    1 * 125 mL - SFr. 65.90
    SFr. 65.90
    1 * 125 mL
       SFr. 65.90
  • 786-567
    Gram negative lysis buffer
    Gram negative lysis buffer
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - SFr. 104.00
    SFr. 104.00
    1 * 250 mL
       SFr. 104.00
  • 786-579
    Gram negative lysis buffer
    Gram negative lysis buffer
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 207.00
    SFr. 207.00
    1 * 500 mL
       SFr. 207.00
  • 786-580
    Gram negative lysis buffer
    Gram negative lysis buffer
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 397.00
    SFr. 397.00
    1 * 1 L
       SFr. 397.00