Protein solubilisation buffer
FOCUS™ protein solubilisation buffer (PSB) with Diluent III, 50 mL
Catalog # 786-PSB
Supplier: G-Biosciences
Jump to:
Product Details & Documents
FOCUS™ protein solubilisation buffer is a dry, urea-based buffer for protein solubilisation. It is a pre-mixed formulation of urea, thiourea, CHAPS and non detergent sulphobetaine for maximum solubilising strength.
- Convenient and simple to use - simply hydrate and use
- Prevents urea induced protein carbamylation
- Prevents waste of unused reagents
Informazioni per l'ordine: Supplied with specific dilution buffer for optimal rehydration and protein solubilisation conditions.
Documents
786-PSB_protocol
Manuale
Specifications
Specifications
- Pk:50 mL
- Description:FOCUS™ protein solubilisation buffer (PSB) with Diluent III
Specifications
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..