Calcium sulphate dihydrate 98.0-102.0%, powder, BAKER ANALYZED® ACS, J.T.Baker®
1452-01
: Avantor
:
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
- Meets ACS Reagent Chemical Requirements:98.0 - 102.0 %
- For Laboratory,Research,or Manufacturing Use:98.0 - 102.0 %
- Assay (CaSO₄·2H₂O) (by EDTA titrn):98.0 - 102.0 %
- Carbonate (CO₃):Passes Test
- Chloride (Cl):≤0.002 %
- Heavy Metals (as Pb):≤0.002 %
- Insoluble in Dilute HCl:≤0.02 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Magnesium (Mg):≤0.02 %
- Nitrate (NO₃):Passes Test
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.02 %
- Strontium (Sr):≤0.05 %
:
:172.17 g/mol
:13,01711
:2.32 g/cm³ (20 °C)
:Ambient
:231-900-3
:10101-41-4
:CaSO₄·2H₂O
:MFCD00149625