Orders LinkContactUsLinkComponent
Stirring rods
Supplier:   HAMMACHER KARL
Stirring rods
Supplier:  HAMMACHER KARL
Certificates
Stirring rods
2 Options Available Below
Product Details & Documents
Stirring rods, made from WIRONIT steel. Choice of rods with straight or angled end.
  • Rust resistant
  • Anti-magnetic
Product Family Options
Product Information
  • Material
    Ø
    Length
    Availability
    Price
  • HAMMHWN070-12
    Straight stirring rod
    Straight stirring rod
    Steel
    2 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.70
    SFr. 12.70
    1 * 1 ST
       SFr. 12.70
  • HAMMHWN071-19
    Angled stirring rod
    Angled stirring rod
    Steel
    4 mm
    194 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 23.70
    SFr. 23.70
    1 * 1 ST
       SFr. 23.70