Order Entry
Switzerland
ContactUsLinkComponent
Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (20 mesh), J.T.Baker®
Irritant
Health
Environment
Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (20 mesh), J.T.Baker®
Catalog # 4244-07
Supplier:  Avantor
CAS Number:  
undefined
Irritant
Health
Environment
Zinc ≥99.8% (by EDTA titration), granular, BAKER ANALYZED® ACS (20 mesh), J.T.Baker®
Catalog # 4244-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    PE pail, screw top, 2 gal.
  • Pk:
    12 kg
  • Seq:
    4187

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • Assay (Zn) (by EDTA titrn):
    ≥99.8 %
  • Iron (Fe):
    ≤0.01 %
  • Lead (Pb):
    ≤0.01 %
  • Suitability for Arsenic Determination:
    Passes Test
  • Trace Impurities - Arsenic (As):
    ≤0.100 ppm
  • Mesh - On U.S. No. 16 Sieve:
    ≤5 %
  • Mesh - Thru U.S. No. 35 Sieve:
    ≤30 %
  • Meets Reagent Specifications for testing USP/NF monographs:

Specification Test Results

About this item

 :
 :65.39 g/mol
Merck Index:12,10255
Boiling Pt:908 °C (1013 hPa)
Melting Pt:420 °C
Density:7.14 g/cm³ (20 °C)
EINECS:231-175-3
 :GHS07
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:7440-66-6
Formula:Zn
MDL Number:MFCD00011291