Order Entry
Switzerland
ContactUsLinkComponent
Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Irritant
Health
Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Health
Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
    -
  • For Laboratory,Research,or Manufacturing Use:
    = 99.5 %
  • Assay ((CH₃)₂CO) (by GC, corrected for water):
    ≥99.5 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤0.001 %
  • Solubility in H₂O:
    Passes Test
  • Titrable Acid (meq/g):
    ≤0.0003
  • Titrable Base (meq/g):
    ≤0.0006
  • Aldehydes (as HCHO):
    ≤0.002 %
  • Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):
    ≤0.05 %
  • Methanol (CH₃OH)(by GC):
    ≤0.05 %
  • Heavy Metals (as Pb):
    ≤1 ppm
  • Substances Reducing Permanganate:
    Passes Test
  • Water (H₂O):
    ≤0.5 %
  • Copper (Cu):
    ≤0.1 ppm
  • Iron (Fe):
    ≤0.1 ppm
  • Nickel (Ni):
    ≤0.1 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together