Diammonium hydrogen orthophosphate ≥98.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Anbieter: Avantor
Zertifikate
Über diesen Artikel
Synonyme: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
CAS-Nummer:7783-28-0
:
Formel:(NH₄)₂HPO₄
MDL-Nummer:MFCD00010891
:132.06 g/mol
Merck-Index:13,00546
Schmelzender Platingehalt:185 °C
Dichte:1.62 g/cm³ (20 °C)
EINECS:231-987-8
ERREICHEN:01-2119490974-22
:GHS07
Lagertemperatur:Ambient
Technische Daten
- Environmentally Preferable:
Technische Daten
Ergebnisse des Spezifikationstests
- Meets ACS Reagent Chemical Requirements:≥98.0%
- For Laboratory,Research,or Manufacturing Use:7.7 - 8.1
- Assay ((NH₄)₂HPO₄) (by acidimetry):≥98.0 %
- pH of 5% Solution at 25°C:7.7 - 8.1
- Insoluble Matter:≤0.005 %
- Chloride (Cl):≤0.001 %
- Nitrate (NO₃):≤0.003 %
- Sulfate (SO₄):≤0.01 %
- Heavy Metals (as Pb):≤0.001 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Calcium (Ca):≤0.001 %
- Magnesium (Mg):≤0.0005 %
- Potassium (K):≤0.005 %
- Sodium (Na):≤0.005 %
Ergebnisse des Spezifikationstests
Frequently Bought Together