Orders LinkContactUsLinkComponent
Handheld spectroscopes
Handheld spectroscopes
Supplier:   A.KRÜSS Optronic
Handheld spectroscopes
Supplier:  A.KRÜSS Optronic
Certificates
Handheld spectroscopes
2 Options Available Below
Product Details & Documents
These handheld spectroscopes are designed for checking the line spectrum of gas discharge lamps as well as individual and comparative observations and spectral examinations.
  • Variable slit and angular dispersion (C-F = 7°)
  • Linear disperson of 60 mm for both models
  • Manual option with grating
Used in education for qualitative analysis and measurement of emission and absorption spectra.
Product Family Options
Product Information
  • Model
    Range
    Availability
    Price
  • KRUE1501
    Handheld spectroscope
    Handheld spectroscope
    1501
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • KRUE1504
    Handheld spectroscope
    Handheld spectroscope
    1504
    400 − 750 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Scale illumination
    Scale illumination
    KRUE15081
    Log in for availability
    1 * 1 ST - 164.00 SFr.
Product Information
  • Catalog #
    Availability
    Price
  • Stand for handheld spectroscopes
    Stand for handheld spectroscopes
    KRUE1510
    Log in for availability
    1 * 1 ST - 173.00 SFr.