ContactUsLinkComponent
Glove boxes, stainless steel combination, Protector®
Glove boxes, stainless steel combination, Protector®
Supplier:  LABCONCO
Certificates
undefined
Glove boxes, stainless steel combination, Protector®
Supplier:  LABCONCO
Specifications
  • Catalog No:
  • LBCP5080172
  • LBCP5080167
  • LBCP5080165
  • LBCP5080170
  • Model:
  • Protector® (double)
  • Protector® (double)
  • Protector®
  • Protector®
  • Observation window (W×H):
  • 2× 910×700 mm
  • 2× 910×700 mm
  • 910×700 mm
  • 910×700 mm
  • W×D×H ext.:
  • 2766×874×1161 mm
  • 2766×874×1161 mm
  • 1613×874×1161 mm
  • 1613×874×1161 mm
  • Weight:
  • 590 kg
  • 590 kg
  • 295 kg
  • 295 kg