Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Chromatography papers

Chromatography papers

Supplier:   Ahlstrom
Chromatography papers
Supplier:  Ahlstrom
Certificates
Chromatography papers

15 Options Available Below

Product Details & Documents

Ahlstrom-Munksjö offers a full range of high quality cotton papers with high absorbency - ideal for chromatographic analysis and blotting methods with gel (Southern, Northern and Western Blots). Ahlstrom-Munksjö manufactures, both in Europe and in the US, absorbent papers made of the highest purity cotton linters. These media do not contain additive of any kind, thus preventing any contamination during the transfer steps.

  • High purity
  • Uniform absorption
  • High intra and inter-lot consistency
  • Wide range of thickness and absorption levels
  • Customised sizing and packaging

Ahlstrom-Munksjö’s absorbent papers are produced with a wide range of thickness and absorption levels for optimal wicking capabilities, drying and blotting in any application. Each media is specifically designed to ensure high quality and consistency. Close surveillance in tightly controlled conditions ensures the highest uniformity, consistency and smoothness of the sheets in all grades. This contributes to clean and even transfers during blotting.

Applications: Chromatography, electrophoresis, Northern, Western and Southern blots, antibiotic discs, seed germination testing.

All references are available in customised sizing and packaging to meet customer needs and industry standards.

Specifications for Product Family

Specifications

  • Catalog No:
  • 514-1182
  • BINZ143124
  • 514-1187
  • 514-1188
  • 514-1189
  • 514-1190
  • BINZ143203
  • 514-1191
  • BINZ143204
  • 514-1192
  • BINZ143225
  • 514-1129
  • BINZ143230
  • BINZ143229
  • 514-1193
  • Grade:
  • FN 1
  • FN 3
  • FN 3
  • FN 4
  • FN 7
  • FN 8
  • FN 100
  • FN 100
  • FN 100
  • FN 100
  • FN 100
  • FN 100
  • FN 100
  • FN 100
  • FN 100
  • Thickness:
  • 0,19 mm
  • 0,19 mm
  • 0,19 mm
  • 0,24 mm
  • 0,32 mm
  • 0,55 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • 0,35 mm
  • Weight:
  • 90 g/m²
  • 90 g/m²
  • 90 g/m²
  • 125 g/m²
  • 150 g/m²
  • 280 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • 195 g/m²
  • Capillary rise:
  • 145 mm/30 min
  • 93 mm/30 min
  • 93 mm/30 min
  • 93 mm/30 min
  • 145 mm/30 min
  • 170 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min
  • 115 mm/30 min

Specifications

Product Family Options

Grade FN 1

Product Information

  • Size
    Availability
    Price
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 271.00
    SFr. 271.00
    1 * 100 ST
       SFr. 271.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1182
    Grade
    FN 1
    Thickness
    0,19 mm
    Weight
    90 g/m²
    Capillary rise
    145 mm/30 min

Grade FN 3

Product Information

  • Size
    Availability
    Price
  • Sheets
    100×300 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 55.20
    SFr. 55.20
    1 * 100 ST
       SFr. 55.20

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143124
    Grade
    FN 3
    Thickness
    0,19 mm
    Weight
    90 g/m²
    Capillary rise
    93 mm/30 min
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 213.00
    SFr. 213.00
    1 * 100 ST
       SFr. 213.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1187
    Grade
    FN 3
    Thickness
    0,19 mm
    Weight
    90 g/m²
    Capillary rise
    93 mm/30 min

Grade FN 4

Product Information

  • Size
    Availability
    Price
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 445.00
    SFr. 445.00
    1 * 100 ST
       SFr. 445.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1188
    Grade
    FN 4
    Thickness
    0,24 mm
    Weight
    125 g/m²
    Capillary rise
    93 mm/30 min

Grade FN 7

Product Information

  • Size
    Availability
    Price
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 221.00
    SFr. 221.00
    1 * 50 ST
       SFr. 221.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1189
    Grade
    FN 7
    Thickness
    0,32 mm
    Weight
    150 g/m²
    Capillary rise
    145 mm/30 min

Grade FN 8

Product Information

  • Size
    Availability
    Price
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 328.00
    SFr. 328.00
    1 * 50 ST
       SFr. 328.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1190
    Grade
    FN 8
    Thickness
    0,55 mm
    Weight
    280 g/m²
    Capillary rise
    170 mm/30 min

Grade FN 100

Product Information

  • Size
    Availability
    Price
  • Sheets
    190×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 297.00
    SFr. 297.00
    1 * 1 ST
       SFr. 297.00

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143203
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    200×200 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 84.00
    SFr. 84.00
    1 * 100 ST
       SFr. 84.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1191
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    230×100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 361.00
    SFr. 361.00
    1 * 1 ST
       SFr. 361.00

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143204
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    350×450 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 193.00
    SFr. 193.00
    1 * 100 ST
       SFr. 193.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1192
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    460×570 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 236.00
    SFr. 236.00
    1 * 50 ST
       SFr. 236.00

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143225
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    460×570 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 424.00
    SFr. 424.00
    1 * 100 ST
       SFr. 424.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1129
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    580×600 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 313.00
    SFr. 313.00
    1 * 50 ST
       SFr. 313.00

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143230
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    580×680 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - SFr. 297.00
    SFr. 297.00
    1 * 50 ST
       SFr. 297.00

    Specifications:
    More
    Specifications
    Cat. No.
    BINZ143229
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
  • Sheets
    580×680 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - SFr. 384.00
    SFr. 384.00
    1 * 100 ST
       SFr. 384.00

    Specifications:
    More
    Specifications
    Cat. No.
    514-1193
    Grade
    FN 100
    Thickness
    0,35 mm
    Weight
    195 g/m²
    Capillary rise
    115 mm/30 min
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.