Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Electrodes
pH Electrodes
Accessories for digital pH electrodes, InLab® smart pro-ISM
Accessories for digital pH electrodes, InLab® smart pro-ISM
METT30027776
:
Mettler - Toledo
Accessories for digital pH electrodes, InLab® smart pro-ISM
METT30027776
:
Mettler - Toledo
Description:
Cable InLab Digital K8SD to Mini-LTW, 1,2 m, for connecting the InLab® Smart Pro-ISM electrode to SevenExcellence™ and SevenCompact™ meters
Pk:
1