Orders LinkContactUsLinkComponent
Foxp3/transcription factor staining buffer
Foxp3/transcription factor staining buffer
Supplier:   Tonbo Biosciences
Foxp3/transcription factor staining buffer
Supplier:  Tonbo Biosciences
Certificates
Foxp3/transcription factor staining buffer
Foxp3/transcription factor staining buffer
3 Options Available Below
Product Details & Documents
Foxp3/transcription factor staining buffer contains specially formulated buffers and solutions for optimal resolution and low background in your analysis of nuclear antigens by flow cytometry. This complete kit contains three components for use in these staining protocols for detection of nuclear antigens such as Foxp3 and ROR gamma. Foxp3/transcription factor fix/perm concentrate (4x) 50 ml. A concentrated solution which, when diluted with Foxp3/transcription factor fix/perm diluent (1x) provides best results in protocols for intranuclear staining of transcription factors using fluorescently conjugated antibodies.
Ordering information: Includes (1) Foxp3/transcription factor fix/Perm Concentrate (4x) 50 ml, a concentrated solution, which when diluted with Foxp3/transcription factor fix/perm diluent (1x) provides best results in protocols for intranuclear staining of transcription factors using fluorescently conjugated antibodies; (2) Foxp3/transcription factor fix/perm diluent (1x) 160 ml intended for use as a diluent for Foxp3/transcription factor fix/perm concentrate (4x); (3) flow cytometry perm buffer (10x) 150 ml provided as a concentrate, which when diluted with distilled water to a 1x solution provides best results in intracellular staining protocols for cytokines and other cytoplasmic antigens, by maintaining membrane permeabilization throughout staining and wash steps.
Product Family Options
Product Information
  • Pk
    Availability
    Price
  • TNB-0607-KIT
    Foxp3/transcription factor staining buffer kit, mouse and human reactivity
    Foxp3/transcription factor staining buffer kit, mouse and human reactivity
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - SFr. 132.00
    SFr. 132.00
    1 * 1 KIT
       SFr. 132.00
  • TNB-1020-L050
    Foxp3/transcription factor fix/perm concentrate (4x), mouse and human reactivity
    Foxp3/transcription factor fix/perm concentrate (4x), mouse and human reactivity
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - SFr. 70.10
    SFr. 70.10
    1 * 50 mL
       SFr. 70.10
  • TNB-1022-L160
    Foxp3/transcription factor fix/perm diluent (1x), mouse and human reactivity
    Foxp3/transcription factor fix/perm diluent (1x), mouse and human reactivity
    160 mL
    Log in for availability
    Log in for availability
    1 * 160 mL
    1 * 160 mL - SFr. 27.20
    SFr. 27.20
    1 * 160 mL
       SFr. 27.20