L-Lysine, anhydrous 98.5-102.0%, Bioreagent for biopharmaceutical production
42307-1KG
:
- Pk:1 kg
- Seq:4024
- Assay (on dried substance):98.5 - 102.0 %
- Appearance:Crystalline powder
- Colour:White to off white
- Cell culture toxicity:Passes test
- DNases:Not detected
- Identification by IR:Conforms to structure
- Proteases:Not detected
- RNases:Not detected
- Solubility (10 %):Clear.complete.yellowish
- Spec. opt. rot.(25°C; 8 %; HCl 6N;dried):25.5 - 26.7 °
- Heavy metals (as Pb):Max. 15 ppm
- Ignition residue:Max. 0.1 %
- Water:Max. 2.0 %
- SO₄ (Sulphate):Max. 0.03 %
- Fe (Iron):Max. 30 ppm
- Bioburden:Max. 100 CFU/g
- Endotoxin (1 %) (EU/mg):Max. 0.1 EU/mg
:
:146.19 g/mol
:13,05656
:290 °C (1013 hPa)
:196 °C
:200-294-2
:56-87-1
:C₆H₁₄N₂O₂
:MFCD00064433