ContactUsLinkComponent
Overalls, PP
Overalls, PP
Catalog # 113-8104
undefined
Overalls, PP
Catalog # 113-8104
Specifications
  • Clothing Size:
    One size
  • Garment application:
    General Purpose,
    Laboratory,
    Industrial,
    Food
  • Pk:
    50
  • Weight:
    52 g/m²
Specifications
About this item
These disposable overalls made from non woven PP provide protection against dust and dirt.
  • With hood
  • Zip closure
  • Elasticated wrist and ankles
  • Single bagged
Suitable for use in low risk areas, such as food processing, manufacturing and hygiene areas. Ideal for visitors.
Weight: 52 g/m²
*One size fits all