Transfer tanks, Transphor™
Catalog # TE22
Anbieter: Cytiva
Technische Daten
- Model:TE22
- Description:Mini transfer tank, Transphor™
- Buffer volume:1000 ml
- Gel size W×L:90×100 mm
- Pk:1
Technische Daten
Über diesen Artikel
Transphor™ units use transverse electrophoresis to transfer proteins from polyacrylamide gels onto nitrocellulose and PVDF membranes. The optional Transphor™ power lid delivers up to 1,5 A and 100 V in constant current mode.
- Performs complete and even transfers of proteins
- Transfers four 15×21 cm gels or up to sixteen 7×10 cm gels at one time
- Cassettes load easily with even contact between gel and transfer membrane
- Sturdy electrode panels lock safely in place automatically