YOUTILITY® bottles, amber, glass, with screw cap, DURAN®
Anbieter: DWK Life Sciences
Zertifikate
Über diesen Artikel
DURAN®, Borosilicate 3.3 glass, amber.
- Amber coloured to protect light-sensitive materials
- Specially shaped grip zones on the bottles for safer and easier handling
- Ergonomic GL 45 screw cap made from food grade polypropylene
- Shortened threads on bottle and cap for faster opening
- Dedicated accessories for easy and simple identification and personalisation
- Glass Type I according to USP, EP and JP
- Amber glass meets USP 671 and Eur. Ph. 3.2.1. for protection against UV light
The DURAN® YOUTILITY bottle represents a new generation of glass laboratory bottles for use in a wide and diverse range of applications. The innovations include improved ergonomic bottle shape and extend to dedicated accessories that incorporate many innovative features to improve handling, sample identification, and ease of use.
With printed Retrace Code (batch identification).
Frequently Bought Together